Assay Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514839.2 | XP_003514887.1 | 6 - 7 | 504 | 73 | |
| XM_007654883.1 | XP_007653073.1 | 6 - 7 | 720 | 73 | ||
| XM_007654884.1 | XP_007653074.1 | 7 - 8 | 859 | 73 | ||
| XM_007654885.1 | XP_007653075.1 | 6 - 7 | 505 | 73 | ||
| GenBank mRNA | JI882703.1 | - | 2 - 3 | 198 | 73 | |
| JP062258.1 | - | 2 - 3 | 196 | 73 | ||
More Information
Set Membership: |
|

Ccdc101
Genomic Map