Assay Details
Gene Symbol: Mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503059.2 | XP_003503107.1 | 2 - 3 | 828 | 63 | |
XM_007643682.1 | XP_007641872.1 | 2 - 3 | 828 | 63 | ||
GenBank mRNA | JP061042.1 | - | 2 - 3 | 404 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
