Assay Details
| Gene Symbol: LOC100752832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506690.2 | XP_003506738.2 | 11 - 12 | 1806 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100752832| Gene Symbol: LOC100752832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506690.2 | XP_003506738.2 | 11 - 12 | 1806 | 58 | |
Set Membership: |
|