Assay Details
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509537.2 | XP_003509585.1 | 5 - 6 | 633 | 71 | |
GenBank mRNA | JI879283.1 | - | 5 - 6 | 466 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
