Assay Details
| Gene Symbol: Lgals1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510270.2 | XP_003510318.1 | 2 - 3 | 219 | 99 | |
| GenBank mRNA | JI882569.1 | - | 2 - 3 | 145 | 99 | |
| JP064756.1 | - | 2 - 3 | 145 | 99 | ||
More Information
Set Membership: |
|

Lgals1
Genomic Map