Assay Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510478.2 | XP_003510526.1 | 8 - 9 | 1342 | 68 | |
| XM_003510479.2 | XP_003510527.1 | 8 - 9 | 1342 | 68 | ||
| GenBank mRNA | JI863029.1 | - | 6 - 7 | 934 | 68 | |
| JP039573.1 | - | 6 - 7 | 934 | 68 | ||
More Information
Set Membership: |
|

Thra
Genomic Map