Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650187.1 | XP_007648377.1 | 5 - 6 | 735 | 72 | |
GenBank mRNA | JI882968.1 | - | 2 - 3 | 319 | 72 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650187.1 | XP_007648377.1 | 5 - 6 | 735 | 72 | |
GenBank mRNA | JI882968.1 | - | 2 - 3 | 319 | 72 |