Assay Details
| Gene Symbol: Xirp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646436.1 | XP_007644626.1 | 2 - 3 | 5253 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xirp1| Gene Symbol: Xirp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646436.1 | XP_007644626.1 | 2 - 3 | 5253 | 98 | |
Set Membership: |
|