Assay Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003494969.2 | XP_003495017.1 | 3 - 4 | 1346 | 60 | |
| GenBank mRNA | JI869908.1 | - | 1 - 2 | 132 | 60 | |
| JP038893.1 | - | 1 - 2 | 132 | 60 | ||
More Information
Set Membership: |
|

Dusp10
Genomic Map