Assay Details
Gene Symbol: Lmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498883.1 | XP_003498931.1 | 1 - 2 | 267 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
