Assay Details
Gene Symbol: Sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500096.2 | XP_003500144.1 | 2 - 3 | 191 | 76 | |
GenBank mRNA | JI882194.1 | - | 2 - 3 | 187 | 76 | |
JP065739.1 | - | 2 - 3 | 187 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
