Assay Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504631.2 | XP_003504679.2 | 4 - 5 | 548 | 67 | |
| XM_007645140.1 | XP_007643330.1 | 3 - 4 | 368 | 67 | ||
More Information
Set Membership: |
|

Hsd17b1
Genomic Map