Assay Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511492.2 | XP_003511540.1 | 11 - 12 | 1249 | 65 | |
| XM_007652036.1 | XP_007650226.1 | 11 - 12 | 1109 | 65 | ||
| GenBank mRNA | JP061719.1 | - | 11 - 12 | 1231 | 65 | |
More Information
Set Membership: |
|

Csnk2a1
Genomic Map