Assay Details
| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513692.2 | XP_003513740.1 | 2 - 3 | 422 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alx3| Gene Symbol: Alx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513692.2 | XP_003513740.1 | 2 - 3 | 422 | 63 | |
Set Membership: |
|