Assay Details
Gene Symbol: Msantd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514725.2 | XP_003514773.1 | 3 - 4 | 552 | 104 | |
XM_007654777.1 | XP_007652967.1 | 2 - 3 | 4228 | 104 | ||
XM_007654778.1 | XP_007652968.1 | 2 - 3 | 4057 | 104 | ||
XM_007654779.1 | XP_007652969.1 | 3 - 4 | 4138 | 104 | ||
XM_007654780.1 | XP_007652970.1 | 3 - 4 | 3854 | 104 | ||
XM_007654781.1 | XP_007652971.1 | 3 - 4 | 4156 | 104 | ||
XM_007654782.1 | XP_007652972.1 | 3 - 4 | 993 | 104 | ||
GenBank mRNA | JI867512.1 | - | 1 - 2 | 103 | 104 | |
JP049265.1 | - | 1 - 2 | 103 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
