Assay Details
| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651134.1 | XP_007649324.1 | 2 - 3 | 1560 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pram1| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651134.1 | XP_007649324.1 | 2 - 3 | 1560 | 71 | |
Set Membership: |
|