Assay Details
| Gene Symbol: Ascl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509905.1 | XP_003509953.1 | 1 | 416 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ascl3| Gene Symbol: Ascl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509905.1 | XP_003509953.1 | 1 | 416 | 65 | |
Set Membership: |
|