Assay Details
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513636.2 | XP_003513684.1 | 2 - 3 | 254 | 93 | |
GenBank mRNA | JI880732.1 | - | 2 - 3 | 228 | 93 | |
JP051574.1 | - | 2 - 3 | 228 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
