Assay Details
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643318.1 | XP_007641508.1 | 5 - 6 | 1274 | 64 | |
XM_007643319.1 | XP_007641509.1 | 6 - 7 | 1396 | 64 | ||
XM_007643320.1 | XP_007641510.1 | 5 - 6 | 997 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
