Assay Details
| Gene Symbol: Dact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644579.1 | XP_007642769.1 | 3 - 4 | 488 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dact2| Gene Symbol: Dact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644579.1 | XP_007642769.1 | 3 - 4 | 488 | 95 | |
Set Membership: |
|