Assay Details
| Gene Symbol: Vsig10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645108.1 | XP_007643298.1 | 3 - 4 | 830 | 67 | |
| XM_007645116.1 | XP_007643306.1 | 3 - 4 | 830 | 67 | ||
More Information
Set Membership: |
|

Vsig10
Genomic Map