Assay Details
| Gene Symbol: LOC100769818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495960.2 | XP_003496008.1 | 2 - 3 | 604 | 79 | |
| XM_003495961.2 | XP_003496009.1 | 2 - 3 | 609 | 79 | ||
More Information
Set Membership: |
|

LOC100769818
Genomic Map