Assay Details
| Gene Symbol: LOC100769818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495961.2 | XP_003496009.1 | 19 - 20 | 2149 | 59 | |
| GenBank mRNA | JI871159.1 | - | 3 - 4 | 291 | 59 | |
More Information
Set Membership: |
|

LOC100769818
Genomic Map