Assay Details
| Gene Symbol: Btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499992.2 | XP_003500040.1 | 2 - 3 | 380 | 70 | |
| GenBank mRNA | JI895827.1 | - | 2 - 3 | 380 | 70 | |
| JP038144.1 | - | 2 - 3 | 378 | 70 | ||
More Information
Set Membership: |
|

Btg3
Genomic Map