Assay Details
Gene Symbol: Zc2hc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502769.2 | XP_003502817.1 | 2 - 3 | 1439 | 95 | |
XM_007643389.1 | XP_007641579.1 | 2 - 3 | 2112 | 95 | ||
XM_007643390.1 | XP_007641580.1 | 2 - 3 | 2116 | 95 | ||
XM_007643391.1 | XP_007641581.1 | 2 - 3 | 1640 | 95 | ||
XM_007643392.1 | XP_007641582.1 | 2 - 3 | 1606 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
