Assay Details
| Gene Symbol: Hoxc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507188.2 | XP_003507236.1 | 1 - 2 | 3097 | 89 | |
| XM_007647747.1 | XP_007645937.1 | 2 - 3 | 380 | 89 | ||
| XM_007647748.1 | XP_007645938.1 | 3 - 4 | 450 | 89 | ||
| XM_007647749.1 | XP_007645939.1 | 2 - 3 | 1166 | 89 | ||
| GenBank mRNA | JI878021.1 | - | 1 - 2 | 106 | 89 | |
More Information
Set Membership: |
|

Hoxc6
Genomic Map