Assay Details
Gene Symbol: Wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503034.1 | XP_003503082.1 | 2 - 3 | 529 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
