Assay Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509683.2 | XP_003509731.1 | 6 - 7 | 882 | 57 | |
| XM_007650321.1 | XP_007648511.1 | 5 - 6 | 813 | 57 | ||
| GenBank mRNA | JI882811.1 | - | 6 - 7 | 832 | 57 | |
| JP039633.1 | - | 6 - 7 | 832 | 57 | ||
More Information
Set Membership: |
|

Arpc1a
Genomic Map