Assay Details
Gene Symbol: Znf483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651197.1 | XP_007649387.1 | 4 - 5 | 830 | 82 | |
XM_007651198.1 | XP_007649388.1 | 4 - 5 | 830 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
