Assay Details
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496862.2 | XP_003496910.1 | 1 - 2 | 163 | 60 | |
GenBank mRNA | JI882640.1 | - | 1 - 2 | 155 | 60 | |
JP043144.1 | - | 1 - 2 | 155 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
