Assay Details
Gene Symbol: Msantd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505536.2 | XP_003505584.1 | 2 - 3 | 696 | 77 | |
XM_007646031.1 | XP_007644221.1 | 2 - 3 | 682 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
