Assay Details
| Gene Symbol: LOC100760251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514130.2 | XP_003514178.2 | 3 - 4 | 252 | 72 | |
| XM_007654324.1 | XP_007652514.1 | 2 - 3 | 201 | 72 | ||
| GenBank mRNA | JI868038.1 | - | 3 - 4 | 486 | 72 | |
More Information
Set Membership: |
|

LOC100760251
Genomic Map