Assay Details
Gene Symbol: Kifc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651409.1 | XP_007649599.1 | 7 - 8 | 1629 | 62 | |
XM_007651411.1 | XP_007649601.1 | 7 - 8 | 1521 | 62 | ||
GenBank mRNA | JI890514.1 | - | 1 - 2 | 154 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
