Assay Details
Gene Symbol: LOC100762730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645325.1 | XP_007643515.1 | 2 - 3 | 249 | 110 | |
XM_007645326.1 | XP_007643516.1 | 2 - 3 | 246 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
