Assay Details
Gene Symbol: Mrpl51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496046.2 | XP_003496094.1 | 2 - 3 | 374 | 70 | |
GenBank mRNA | JI881554.1 | - | 2 - 3 | 265 | 70 | |
JP048559.1 | - | 2 - 3 | 265 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
