Assay Details
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502055.2 | XP_003502103.1 | 3 - 4 | 1679 | 66 | |
XM_003502057.2 | XP_003502105.1 | 3 - 4 | 1649 | 66 | ||
XM_007642674.1 | XP_007640864.1 | 4 - 5 | 965 | 66 | ||
XM_007642675.1 | XP_007640865.1 | 3 - 4 | 677 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
