Assay Details
| Gene Symbol: Sdc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504581.2 | XP_003504629.1 | 3 - 4 | 301 | 74 | |
| GenBank mRNA | JI865108.1 | - | 3 - 4 | 279 | 74 | |
| JP062945.1 | - | 3 - 4 | 283 | 74 | ||
More Information
Set Membership: |
|

Sdc4
Genomic Map