Assay Details
Gene Symbol: Scara3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505164.2 | XP_003505212.1 | 1 - 2 | 187 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
