Assay Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510739.2 | XP_003510787.1 | 3 - 4 | 393 | 62 | |
| XM_007651336.1 | XP_007649526.1 | 3 - 4 | 393 | 62 | ||
| XM_007651337.1 | XP_007649527.1 | 2 - 3 | 165 | 62 | ||
| XM_007651338.1 | XP_007649528.1 | 2 - 3 | 165 | 62 | ||
| GenBank mRNA | JI890042.1 | - | 1 - 2 | 90 | 62 | |
| JP042137.1 | - | 1 - 2 | 90 | 62 | ||
More Information
Set Membership: |
|

Fkbp11
Genomic Map