Assay Details
Gene Symbol: LOC100766941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513684.2 | XP_003513732.1 | 1 - 2 | 153 | 77 | |
XM_007653935.1 | XP_007652125.1 | 1 - 2 | 153 | 77 | ||
GenBank mRNA | JI874762.1 | - | 1 - 2 | 142 | 77 | |
JP055976.1 | - | 1 - 2 | 147 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007653935.1 transcripts.
Set Membership: |
![]() ![]() |
