Assay Details
Gene Symbol: LOC100766941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513684.2 | XP_003513732.1 | 6 - 7 | 587 | 78 | |
XM_007653935.1 | XP_007652125.1 | 7 - 8 | 649 | 78 | ||
XM_007653936.1 | XP_007652126.1 | 3 - 4 | 215 | 78 | ||
XM_007653937.1 | XP_007652127.1 | 5 - 6 | 510 | 78 | ||
GenBank mRNA | JI874762.1 | - | 6 - 7 | 576 | 78 | |
JP055976.1 | - | 6 - 7 | 581 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
