Assay Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642329.1 | XP_007640519.1 | 2 - 3 | 467 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35b2| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642329.1 | XP_007640519.1 | 2 - 3 | 467 | 68 | |
Set Membership: |
|