Assay Details
Gene Symbol: LOC100762617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641838.1 | XP_007640028.1 | 1 - 2 | 878 | 88 | |
XM_007641839.1 | XP_007640029.1 | 2 - 3 | 432 | 88 | ||
XM_007641840.1 | XP_007640030.1 | 1 - 2 | 878 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
