Assay Details
Gene Symbol: Muc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654879.1 | XP_007653069.1 | 6 - 7 | 524 | 76 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Muc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654879.1 | XP_007653069.1 | 6 - 7 | 524 | 76 |