Assay Details
| Gene Symbol: Mapk1ip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500046.2 | XP_003500094.1 | 3 - 4 | 859 | 84 | |
| GenBank mRNA | JP056405.1 | - | 3 - 4 | 855 | 84 | |
More Information
Set Membership: |
|

Mapk1ip1l
Genomic Map