Assay Details
| Gene Symbol: Tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507516.2 | XP_003507564.1 | 4 - 5 | 535 | 76 | |
| GenBank mRNA | JI885443.1 | - | 1 - 2 | 41 | 76 | |
| JP060873.1 | - | 1 - 2 | 41 | 76 | ||
More Information
Set Membership: |
|

Tsfm
Genomic Map