Assay Details
Gene Symbol: Cntf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514488.2 | XP_003514536.1 | 1 - 2 | 1687 | 77 | |
XM_007654607.1 | XP_007652797.1 | 2 - 3 | 326 | 77 | ||
XM_007654608.1 | XP_007652798.1 | 2 - 3 | 222 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
