Assay Details
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514821.2 | XP_003514869.1 | 5 - 6 | 951 | 63 | |
XM_007654875.1 | XP_007653065.1 | 5 - 6 | 914 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
