Assay Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504782.2 | XP_003504830.2 | 3 - 4 | 419 | 61 | |
GenBank mRNA | JI893547.1 | - | 1 - 2 | 97 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
