Assay Details
| Gene Symbol: LOC100774485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506071.1 | XP_003506119.1 | 4 - 5 | 652 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100774485| Gene Symbol: LOC100774485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506071.1 | XP_003506119.1 | 4 - 5 | 652 | 57 | |
Set Membership: |
|